missing translation for 'onlineSavingsMsg'
Learn More

(R)-α-Methylphenylalanine hydrate, 98%, 98% ee, Thermo Scientific™

Catalog Number p-4259028
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
17350-84-4
C10H13NO2·xH2O
197.23
HYOWVAAEQCNGLE-SNVBAGLBSA-N
r-2-amino-2-methyl-3-phenylpropanoic acid, alpha-methyl-d-phenylalanine, alpha-methyl-d-phe, 2r-2-amino-2-methyl-3-phenylpropanoic acid, a-benzyl-d-ala, d-alpha-methyl phenylalanine, h-alpha-me-d-phe-oh, r-2-amimo-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropionic acid, alpha-benzyl-d-ala
7023409
(2R)-2-amino-2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
This item is not returnable. View return policy
17350-84-4
C10H13NO2·xH2O
197.23
HYOWVAAEQCNGLE-SNVBAGLBSA-N
r-2-amino-2-methyl-3-phenylpropanoic acid, alpha-methyl-d-phenylalanine, alpha-methyl-d-phe, 2r-2-amino-2-methyl-3-phenylpropanoic acid, a-benzyl-d-ala, d-alpha-methyl phenylalanine, h-alpha-me-d-phe-oh, r-2-amimo-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropionic acid, alpha-benzyl-d-ala
7023409
(2R)-2-amino-2-methyl-3-phenylpropanoic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point >233°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Specific Rotation 18
Formula Weight 197.23
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (R)-α-Methylphenylalanine hydrate, 98% ee
Show More Show Less
Product Identifier
  • (R)-alpha-Methylphenylalanine hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'