missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-3-Amino-1-BOC-piperidine, 97%, Thermo Scientific Chemicals

Catalog Number p-4228294
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
188111-79-7
C10H20N2O2
200.28
AKQXKEBCONUWCL-MRVPVSSYSA-N
r-1-boc-3-aminopiperidine, r-tert-butyl 3-aminopiperidine-1-carboxylate, r-3-amino-1-n-boc-piperidine, tert-butyl 3r-3-aminopiperidine-1-carboxylate, r-1-boc-3-amino-piperidine, r-3-amino-1-boc-piperidine, s-n-boc-3-aminopiperidine, r-3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3r, r-1-tert-butoxycarbonyl-3-aminopiperidine
1501974
tert-butyl (3R)-3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N
This item is not returnable. View return policy
188111-79-7
C10H20N2O2
200.28
AKQXKEBCONUWCL-MRVPVSSYSA-N
r-1-boc-3-aminopiperidine, r-tert-butyl 3-aminopiperidine-1-carboxylate, r-3-amino-1-n-boc-piperidine, tert-butyl 3r-3-aminopiperidine-1-carboxylate, r-1-boc-3-amino-piperidine, r-3-amino-1-boc-piperidine, s-n-boc-3-aminopiperidine, r-3-amino-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 3r, r-1-tert-butoxycarbonyl-3-aminopiperidine
1501974
tert-butyl (3R)-3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -30
Formula Weight 200.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (R)-(-)-3-Amino-1-BOC-piperidine
Show More Show Less
compliance-icons
Product Identifier
  • (R)-1-BOC-3-Aminopiperidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'