Learn More
Quinoline-6-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 25 g |
Formula Weight | 173.17 |
Percent Purity | 97% |
Chemical Name or Material | Quinoline-6-carboxylic acid |