missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quetiapine hemifumarate

Catalog Number p-7214412
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
111974-72-2
C29H33N3O10S
615.65
MFCD03423782
JLWSQVHJLLYDPX-LVEZLNDCSA-N
bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12
This item is not returnable. View return policy
111974-72-2
C29H33N3O10S
615.65
MFCD03423782
JLWSQVHJLLYDPX-LVEZLNDCSA-N
bis((2E)-but-2-enedioic acid); 2-[2-(4-{2-thia-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-10-yl}piperazin-1-yl)ethoxy]ethan-1-ol
OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OCCOCCN1CCN(CC1)C1=NC2=CC=CC=C2SC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172.0°C to 176.0°C
Color Clear Colorless to Light Yellow
Quantity 5 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in dimethylformamide, slightly soluble in methanol,very slightly soluble, in ether, insoluble in ether and acetonitrile
Formula Weight 441.54
Chemical Name or Material Quetiapine hemifumarate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'