Learn More
Macherey-Nagel™ Quantofix™ Carbonate Hardness Test Strip Kits
Allows for the quick and easy determination of alkalinity or carbonate hardness in water. Macherey Nagel™ Quantofix™ Carbonate Hardness Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Brand: Macherey-Nagel™ 91323
Additional Details : Weight : 0.03600kg
Description
- Ready to use
- Rapid: provides reliable results within 30 seconds
- Excellent for the safe control of water in swimming pools and aquariums
- Precise: colour charts adjusted and checked using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
Bright Green to Blue | |
Determination of Carbonate Hardness | |
100 Pack | |
1 min. |
Strip | |
0, 3, 6, 10, 15, 20°d | |
30°C | |
Semi-qualitative Test Strip |