missing translation for 'onlineSavingsMsg'
Learn More

Binder™ IQ/OQ Qualification Folder

For Use With: KBW 240, 400, 720

Brand:  Binder™ 8012-0877

Additional Details : Weight : 0.83000kg

Product Code. 15621966

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

  • Contains IQ/OQ checklists and unit schematics
  • QM certificate in accordance with ISO 9001

Specifications

Specifications

Qualification folder IQ/OQ
KBW 240, 400, 720
Checklists, unit schematics
Videos
Documents