missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals qPCR and Go SYBR No-ROX Kit
Developed for fast, highly reproducible, real-time PCR and validated on commonly used real-time PCR instruments.
Brand: MP Biomedicals 11EBI03200
Additional Details : Weight : 0.00500kg
Description
- Fast and sensitive.
- Highly reproducible, real-time PCR
- Validated on commonly used real-time PCR instruments.
- Combination of the latest advances in buffer chemistry and enhancers, together with an antibody-mediated hot-start DNA polymerase system.
- Provided as a 2x mastermix containing all the components necessary for real-time PCR, including the SYBR™ Green I dye, dNTPs, stabilizers and enhancers. Only primers and template required.
- Restricted product and can only be purchased by approved account holders.
Specifications
SYBR™ Green I dye, dNTPs, stabilizers and enhancers | |
Gene Expression | |
2000 reactions |