Learn More
Pyrimidine, 99%, Thermo Scientific Chemicals
Catalog Number p-3779533
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
289-95-2
C4H4N2
80.09
MFCD00006059
CZPWVGJYEJSRLH-UHFFFAOYSA-N
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
9260
CHEBI:16898
pyrimidine
C1=CN=CN=C1
289-95-2
C4H4N2
80.09
MFCD00006059
CZPWVGJYEJSRLH-UHFFFAOYSA-N
1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
9260
CHEBI:16898
pyrimidine
C1=CN=CN=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 20°C to 22°C
Color Colorless to Orange
Density 1.0100g/mL
Boiling Point 123°C to 124°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5010 to 1.504
Quantity 5 g
Beilstein 23, 89
Merck Index 15, 8100
Specific Gravity 1.01
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 80.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • Pyrimidine
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement