Learn More
Cole-Parmer™ Plastic Luer Stopcocks and Manifolds
Regulate flow without any leaking using these fittings. Cole-Parmer™ Plastic Luer Stopcocks and Manifolds are available with natural PVDF body and polypropylene insert, or all-black PVDF construction.
Brand: Cole-Parmer™ 31200-70
Additional Details : Weight : 0.00090kg
Description
- Natural PVDF stopcocks available in 2- or 3-way configurations
- Black PVDF stopcocks are 1-way
Specifications
PVDF | |
Tubing Adapter |