Learn More
Masterflex™ PVDF NPT Male Adapter to Hose Barb
Connect fluid paths without the use of tools
Brand: Masterflex™ 30704-16
Additional Details : Weight : 0.80000kg
Description
- Quick and easy assembly without tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
- Chemical resistant
- Available in a variety of sizes
Specifications
Translucent | |
Autoclavable | |
1 29/32 in. | |
1/2 in. | |
1/2 in. | |
Autoclave or Ethylene Oxide |
Barb x NPT(M) Connection | |
PVDF | |
NPT Male Adapter to Hose Barb | |
1/2 in. | |
3/8 in. | |
3/8 in. |