Learn More
Masterflex™ PVDF Barbed NPT Male Pipe Adapter Elbows
Designed for quick and easy assembly without tools
Brand: Masterflex™ 30704-30
Additional Details : Weight : 0.80000kg
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Translucent | |
Autoclavable | |
3/4 in. | |
1/8 in. | |
1/8 in. | |
Autoclave or Ethylene Oxide |
1/8 NPT(M) x 1/8 in. ID | |
PVDF | |
Barbed NPT Male Pipe Adapter Elbows | |
1/8 in. | |
1/8 in. | |
1/8 in. |