Learn More
Dostmann Electronic™ PVC Immersion Probes
With an accuracy of ± 0.3°C, and a temperature range of -200 to + 500°C, this immersion probe is ideal for use in laboratory, scientific, and other research environments.
130.00€ - 409.00€
Product Code | Brand | Length (Metric) | Description | Sensor Types | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Description | Sensor Types | Price | Quantity & Availability | |||
10259461
|
Dostmann Electronic™
6000-1002 |
300 mm | 3 mm dia. | Class B |
139.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||
10688002
|
Dostmann Electronic
6000-1001 |
150 mm | 3 mm dia. | Class B |
130.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||
10331492
|
Dostmann Electronic
6000-1018 |
150 mm | 3mm dia. | Class 1/3 |
200.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||
11852881
|
Dostmann Electronic™
60001774 |
300 mm | 3mm dia. | Class 1/10 |
359.00€
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
11862881
|
Dostmann Electronic™
60001784 |
300 mm | 6mm dia. | Class 1/10 |
409.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
Description
- Accessories for hand-held meters Series P700
- PT100 immersion probe
- Immersion probe Pt100
- -50 to +350°C, Cl. B, 150 x 3mm
- Handheld meter accessory
Specifications
Probe, Immersion | |
PVC |