missing translation for 'onlineSavingsMsg'
Learn More

Prostaglandin A2, TRC

Product Code. 30428782
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428782 100 mg -
30420481 10 mg -
30423302 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428782 Supplier Toronto Research Chemicals Supplier No. TRCP838550100MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13345-50-1
Molecular Formula C20H30O4
Molecular Weight (g/mol) 334.45
Synonym (5Z,13E,15S)-15-Hydroxy-9-oxoprosta-5,10,13-trien-1-oic Acid, 7-[2-(3-Hydroxy-1-octenyl)-5-oxo-3-cyclopenten-1-yl]-5-heptenoic Acid, (+)-Prostaglandin A2, (15S)-PGA2, 15(S)-Hydroxy-9-oxo-5-cis- 10,13-trans-prostatrienoic Acid, 15(S)-Hydroxy-9-oxo-5-cis-10,13-trans-prostenoic Acid, 15(S)-Prostaglandin A2, 15α-Hydroxy-9-oxo-cis-5,10,trans- 13-prostatrienecarboxylic Acid, 15α-Hydroxy-9-oxo-cis-5,10,trans-13-prostatrienoic Acid, 5,6-cis-PGA2, Medullin, NSC 165561, PGA2
IUPAC Name (Z)-7-[(1R,2S)-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopent-3-en-1-yl]hept-5-enoic acid
SMILES CCCCC[C@H](O)\C=C\[C@H]1C=CC(=O)[C@@H]1C\C=C/CCCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H30O4/c1-2-3-6-9-17(21)14-12-16-13-15-19(22)18(16)10-7-4-5-8-11-20(23)24/h4,7,12-18,21H,2-3,5-6,8-11H2,1H3,(H,23,24)/b7-4-,14-12+/t16-,17-,18+/m0/s1
Formula Weight 334.2144
Percent Purity >95%
Chemical Name or Material Prostaglandin A2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.