missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPP1CA (Human) Recombinant Protein
Human PPP1CA full-length ORF ( AAH08010.1, 1 a.a. - 330 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005499-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSDSEKLNLDSIIGRLLEVQGSRPGKNVQLTENEIRGLCLKSREIFLSQPILLELEAPLKICGDIHGQYYDLLRLFEYGGFPPESNYLFLGDYVDRGKQSLETICLLLAYKIKYPENFFLLRGNHECASINRIYGFYDECKRRYNIKLWKTFTDCFNCLPIAAIVDEKIFCCHGGLSPDLQSMEQIRRIMRPTDVPDQGLLCDLLWSDPDKDVQGWGENDRGVSFTFGAEVVAKFLHKHDLDLICRAHQVVEDGYEFFAKRQLVTLFSAPNYCGEFDNAGAMMSVDETLMCSFQILKPADKNKGKYGQFSGLNPGGRPITPPRNSAKAKK
Specifications
AAH08010.1 | |
protein phosphatase 1, catalytic subunit, alpha isoform | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPP1CA | |
GST |
5499 | |
Wheat germ expression system | |
25 μg | |
MGC15877, MGC1674, PP-1A, PPP1A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |