missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Sodium Tartrate Tetrahydrate, 99.0-102.0%, (ACS Reagent Grade), MP Biomedicals
73.57 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Use promo code "23257" to get your promotional price.
Click to view available options
Quantity:
500 g
3 kg
Specifications
Quantity | 500 g |
Assay Percent Range | 99.0-102.0% |
Formula Weight | 282.22 |
Grade | ACS Reagent |
Chemical Name or Material | Potassium Sodium Tartrate Tetrahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'