missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrite, ACS, 96.0% min, Thermo Scientific Chemicals
17.73 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
100 g
500 g
Description
In analytical chemistry (testing for amino acids, cobalt, iodine, urea).Potassium nitrite is used as a food preservative, paint and coating additives and analytical reagent. It is used as a strong oxidizing agent as well as involved in the preparation of heat transfer salts. In addition, it is employed as corrosion inhibitors and anti-scaling agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 350°C (decomposition) |
Physical Form | Solid |
Quantity | 10 g |
Assay Percent Range | 96.0% min |
Solubility Information | Soluble in water,hot alcohol,liquid NH3Soluble in water,alcohol and ammonia. |
Formula Weight | 85.11 |
Grade | ACS Reagent |
Sensitivity | Hygroscopic |
Density | 1.915 g/mL |
Chemical Name or Material | Potassium nitrite, For ACS analysis |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'