missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Potassium Nitrate, ≥99.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent,≥99.0%

Brand:  Honeywell Fluka™ 221295-12KG

Additional Details : CAS : 7757-79-1 Weight : 12.00000kg Transport : UN number : 1486 Chem class : 5.1 Pack group : III


Packaging Drum with inliner
Quantity 12kg

 View more versions of this product

Product Code. 15622110

  • 837.29€ / 12kg


please call +351214253350
Add to basket


Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]

Specifications

Potassium Nitrate
KNO3
UN1486
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.1g/mol
ACS Reagent
12kg
7757-79-1
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
≥99.0%
Drum with inliner
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS