missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution 0.1M (0.1N), (In Ethanol), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
3553
This item is not returnable. View return policy
3553
Quantity 1 L
Packaging HDPE plastic bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide solution 0.1M (0.1N) in alcohol volumetric analysis Standard Volumetric Solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol, Potassiumhydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'