missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Difluoride, purum p.a, ≥99.0% (F), Honeywell™

Catalog Number p-7120704
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
7789-29-9
MFCD00011403
This item is not returnable. View return policy
7789-29-9
MFCD00011403

Purum p.a.,≥99.0% (F)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Boiling Point >180°C
Melting Point 239°C
pH Acidic
Physical Form Solid
Quantity 250 g
Linear Formula KHF2
UN Number UN1811
Solubility Information 392g/L
Formula Weight 78.1g/mol
Percent Purity ≥99.0% (F)
Packaging Poly bottle
Density 2.37g/cm3
Chemical Name or Material Potassium hydrogen difluoride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'