missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Disulfite, puriss. p.a., Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | K2S2O5 |
UN Number | NONH for all modes of transport |
Formula Weight | 222.32g/mol |
Percent Purity | ≥96% (iodometric) |
Grade | puriss. p.a. |
Chemical Name or Material | Potassium Disulfite |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'