missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, ACS Reagent, Honeywell Fluka™

Catalog Number 15621790
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7778-50-9
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7778-50-9
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 500 g
Linear Formula K2Cr2O7
UN Number 3086
Formula Weight 294.18g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Potassium Dichromate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'