missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chlorate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | KClO3 |
UN Number | 1485 |
Formula Weight | 122.55g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Potassium Chlorate, Reag. Ph. Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'