missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide (Beads or Powder/Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
3 kg
Packaging:
Poly Bottle
Specifications
Boiling Point | 1435°C |
Melting Point | 734°C |
Color | White |
pH | 5 to 8.8 |
Physical Form | Solid |
Quantity | 3 kg |
Assay Percent Range | ≥99 % |
Formula Weight | 119 |
Percent Purity | ≥99% |
Grade | Certified ACS |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'