missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Catalog Number p-7120677
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.142
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
This item is not returnable. View return policy
127-08-2
C2H3KO2
98.142
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 292°C
Physical Form Solid
Quantity 100 g
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Solubility Information Soluble in water, 98.2g/L at 20°C
Formula Weight 98.14g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.57g/cu.cm at 25°C
Chemical Name or Material Potassium Acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'