Learn More
Porcelaines Avignon™ Porcelain Mortar
Crush and grind laboratory substances into powders using the Avignon™ Porcelain Mortar. Along with the pestle, it is an integrated laboratory tool in the world of science and medicine.
23.11€ - 264.87€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11882542
|
Porcelaines Avignon™
15014 |
3000 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11758413
|
Porcelaines Avignon™
15010 |
1500 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11832851
|
Porcelaines Avignon™
MOR5+PILPORC3 |
250 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11842851
|
Porcelaines Avignon™
MOR3+PILPORC2 |
150 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11852851
|
Porcelaines Avignon™
MOR1+PILPORC2 |
50 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Sturdy porcelain
- Used with pestle
- Effective for wide variety of laboratory crushing needs
Specifications
Mortar | |
Porcelain | |
Pestle | |
Mortar |