missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POP7 Monoclonal Antibody (3F12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524443
Additional Details : Weight : 0.01000kg
Description
Peptide Sequence: MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINH
POP7 Monoclonal antibody specifically detects POP7 in Human samples. It is validated for ELISA.Specifications
POP7 | |
Monoclonal | |
Unconjugated | |
POP7 | |
0610037N12Rik; hPOP7; POP7; POP7 (processing of precursor, S. cerevisiae) homolog; POP7 homolog, ribonuclease P/MRP subunit; processing of precursor 7, ribonuclease P subunit; processing of precursor 7, ribonuclease P/MRP subunit; ribonuclease P protein subunit p20; Ribonucleases P/MRP protein subunit POP7 homolog; RNaseP protein p20; RPP2; RPP20 | |
Mouse | |
Protein A | |
RUO | |
10248 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
3F12 | |
PBS with no preservative; pH 7.4 | |
O75817 | |
POP7 | |
POP7 (NP_005828.1, 1 a.a. ∽ 80 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |