Learn More
Witeg™ Polypropylene Screw Caps
Catalog Number p-8008791
Click to view available options
Color:
Red
Yellow
Use Witeg™ Polypropylene Screw Caps to keep contents inside.your laboratory bottles. Various colors help you categorize your items.
  • Autoclavable up to 140°C
  • GL 45 Thread
  • Closed
Closure Material Polypropylene
Closure Size GL 25
Closure Type Screw Cap
Product Type Screw Cap
Color Red
Includes 15mm Hole
Material Polypropylene
No. per Pack 10Pack
Quantity 10Pack
Screw Color Red
Screw Material Polypropylene
Closure Color Red
Autoclavable Autoclavable