Learn More
Socorex™ Polypropylene Pipette Tips
Use Socorex™ Polypropylene Pipette Tips when a whole volume range since there is a whole volume range from which to choose. They are available in various colors, are constructed of polypropylene and are autoclavable for cost effectiveness.
Brand:  Socorex™ 309.1000R
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10003511
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Supplied in loose in bags or racks with lids
  • Autoclavable at 121°C / 250°F
  • Compatible with other brands' tips
Specifications
Specifications
Rack
1000μL
PP (Polypropylene)
Universal Fit
Natural
Videos
Documentation
Technical Documents