Learn More
Cole-Parmer™ Polypropylene Male Luer Fitting
Brand:  Cole-Parmer™ 45518-54
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15384732
  • 7.75€ / Pack of 25
please call +351214253350
Add to basket
Specifications
Specifications
Polypropylene
Fits Male Slip Luer x 1/4 in. hose barb
25 Pack
Luer Fitting
Ideal for a variety of applications, including syringes or small tubing