missing translation for 'onlineSavingsMsg'
Learn More

Semadeni™ Polypropylene Flip-Top Boxes

Allows for the safe and secure packing of small items.

Brand:  Semadeni™ 341

Product Code. 12749262

  • 6.00€ / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Practical hinged flip-top container
  • Manufactured from dimensionally stable polypropylene
  • Grey in color
  • Lid and container made from a single block, snap-on cap and integral hinge
  • Ideal for the storage and shipping of small parts
Specifications

Specifications

1.47 L
Gray
Square
Small parts
Storage Box
185 mm
140 mm
133 mm
80 mm
Snap-on Lid with Integral Hinge
Polypropylene
Packaging, Storage
Lid
185 mm
178 mm
140 mm
80 mm
78 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.