Learn More
Macherey-Nagel™ Polygoprep™ 300 to 30 C18
Brand: Macherey-Nagel™ 711720.1000
Additional Details : Weight : 0.01000kg
Specifications
25 to 40 μm | |
POLYGOPREP 300 to 30 | |
1 kg | |
Polygoprep |
300 Å | |
2 to 8 | |
POLYGOPREP C18 | |
Liquid Chromatography |