Learn More
Macherey-Nagel™ Polyester Sheets Polygram SIL G, CEL 300 TLC Micro-Set F1
Brand: Macherey-Nagel™ 814200
Additional Details : Weight : 0.01000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Specifications
Polyester Sheets Polygram SIL G, CEL 300 | |
TLC Micro-Set F1 | |
4 x 8 cm |
1 Ea. | |
1 manual; 50 glass capillaries 1 μL; 50 polyester sheets each of Polygram SIL G/UV254 and CEL 300, 4 x 8 cm; 100 mL each of n-butanol, ninhydrin spray reagent (in ethanol), acetone, 25 % ammonia solution, rubeanic acid spray reagent (in ethanol)*; 50 mL each of 50 % acetic acid, 18 % hydrochloric acid*; 8 mL each of the amino acid test mixture (see above), arginine reference solution and tryptophan reference solution (each aq. soln.); 8 mL each of the heavy metal cation test mixture (see above), Mn(II) reference solution and Cu(II) reference solution (each aq. soln.) |
Safety and Handling
- TLC microset F1
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H318-Causes serious eye damage.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : acetic acid, acetone, ammonia solution,manganese(II) sulphate monohydrate, copper(II) sulphate pent