Learn More
Thermo Scientific™ Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Product Code. 11367111 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Sterile Filter, 0.1μm
Sterile Filter, 0.2μm
Unit Size:
1 unit
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
11367111 Sterile Filter, 0.2μm Each
15950206 Sterile Filter, 0.1μm 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11367111 Supplier Thermo Scientific™ Supplier No. 09.1003
Edge
Sterile filter for use with Barnstead™ water purification systems; can be autoclaved up to 5 times.
Reduce bacteria using the Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems.
Replacement consumable for the following Barnstead water purification systems:
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • LabTower™ EDI
  • LabTower™ TII
  • MicroPure™
  • Smart2Pure™
TRUSTED_SUSTAINABILITY
Specifications
Description Sterile Filter, 0.2μm
For Use With (Equipment) GenPure, GenPure xCAD, LabTower EDI, GenPure Pro
Type Point of Dispense Filter