missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Pneumatic Airgo Crimper

One hand, secure, reproducible crimps/decrimps of 8, 11, 13 and 20mm vials with the push of a button.

Brand:  Fisherbrand™

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 15502300

  • 1435.00€ / Each


Estimated Shipment date
from Supplier 19-07-2018
Add to basket

Description

Description

  • 11mm AIRGO Crimper Low Pressure
  • Pneumatic
  • Steel
  • Crimper
  • Reduces hand strain compared to manual crimper operation
  • Quick and easy removal of aluminum seals with the push of a button
Specifications

Specifications

No
Steel
Aluminum Cap
11mm
Low Pressure
Pneumatic Crimper
Videos