Learn More
Copan Italia™ Single Plastic Chlamydia Transport Swabs
Use Cary-Blair Agar Gel Medium with Copan Italia™ Single Plastic Chlamydia Transport Swabs. These liquid swabs are made with natural fibers with a red plastic handle.
Brand:  Copan Italia™ 132C.USE
Additional Details : Weight : 0.67700kg
Product Code. 11870260
  • 84.00€ / Pack of 50
please call +351214253350
Add to basket
Description
Description
  • For use with Cary-Blair Agar Gel Medium
  • Red Handle
  • Natural Fibers
  • Chlamydia
Specifications
Specifications
Red
50Pack
Yes
10Pack
Natural fibers
Cary-Blair Agar Gel
Swab
Regular
Plastic