Learn More
Copan Italia™ Single Plastic Chlamydia Transport Swabs
Use Cary-Blair Agar Gel Medium with Copan Italia™ Single Plastic Chlamydia Transport Swabs. These liquid swabs are made with natural fibers with a red plastic handle.
Brand: Copan Italia™ 132C.USE
Additional Details : Weight : 0.67700kg
Description
- For use with Cary-Blair Agar Gel Medium
- Red Handle
- Natural Fibers
- Chlamydia
Specifications
| Red | |
| 50Pack | |
| Yes | |
| 10Pack | |
| Natural fibers |
| Cary-Blair Agar Gel | |
| Swab | |
| Regular | |
| Plastic |