Learn More
Thermo Scientific™ Western Blotting Filter Papers, 0.83-mm thick, 8 x 13.5 cm
Pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels to PVDF, nitrocellulose, or other membranes.
59.50€ - 74.00€
Products 7
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
11814131 View Documents Thermo Scientific™
88600
Pierce Western Blotting Filter Paper, 8cm x 10.5cm 100 Sheets 67.25€
Pack of 100 View alternative products
Save Up to 
»
EU Stock 116
Add to basket
10165844 View Documents Thermo Scientific™
88605
Pierce Western Blotting Filter Paper, Extra Thick, 7cm x 8.4cm 50 Sheets 59.50€
Pack of 50 View alternative products
Save Up to 
»
EU Stock 54
Add to basket
10074584 View Documents Thermo Scientific™
88610
Pierce Western Blotting Filter Paper, Extra Thick, 8.5cm x 9cm 50 Sheets 59.50€
Pack of 50 View alternative products
Save Up to 
»
EU Stock 19
Add to basket
11766446 View Documents Thermo Scientific™
88615
Pierce Western Blotting Filter Paper, Extra Thick, 8cm x 13.5cm 50 Sheets 62.00€
Pack of 50 View alternative products
Save Up to 
»
EU Stock 17
Add to basket
10675935 View Documents Thermo Scientific™
88620
Pierce Western Blotting Filter Paper, Extra Thick, 20cm x 20cm 50 Sheets 74.00€
Pack of 50 View alternative products
Save Up to 
»
EU Stock 32
Add to basket
15105214 View Documents Thermo Scientific™
84783
Pierce Western Blotting Filter Paper, 7cm x 8.4cm 100 Sheets 65.00€
100 sheets View alternative products
Save Up to 
»
EU Stock 68
Add to basket
15115214 View Documents Thermo Scientific™
84784
Pierce Western Blotting Filter Paper, 8cm x 13.5cm 100 Sheets 70.75€
100 sheets View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 20-03-2020
Add to basket
Description
Description
Sheets of blotting paper are necessary components of transfer sandwiches and cassettes that typically must be assembled for various kinds of protein or nucleic acid, gel-to-membrane, transfer protocols. High-quality Pierce Western Blotting Filter Papers are available as standard-thickness and extra-thick sheets that are pre-cut for convenience with mini gels.
Highlights:
  • High-quality – clean cotton cellulose fiber paper manufactured with additive-free ultrapure water to eliminate sources of background signal and artifacts
  • Easy to use – pre-cut filter paper sheets in several convenient sizes for use with most mini gel sizes, tank transfer cassettes and semi-dry blotters
  • Two thicknesses – choose standard-thickness paper for traditional procedures or extra-thick filter paper for high capacity blotting or to eliminate using multiple sheets
  • Validated – tested and recommmend for use with protein methods, including the Thermo Scientific Pierce Fast Semi-Dry Blotter (Part No. 88217)