Learn More
Thermo Scientific™ Western Blotting Filter Papers, 0.83-mm thick, 8 x 13.5 cm
Pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels to PVDF, nitrocellulose, or other membranes.
59.50€ - 74.00€
| Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
| 11814131 |
Thermo Scientific™
88600 |
Pierce Western Blotting Filter Paper, 8cm x 10.5cm | 100 Sheets |
67.25€ Pack of 100 |
|
10165844 |
Thermo Scientific™
88605 |
Pierce Western Blotting Filter Paper, Extra Thick, 7cm x 8.4cm | 50 Sheets |
59.50€ Pack of 50 |
|
10074584 |
Thermo Scientific™
88610 |
Pierce Western Blotting Filter Paper, Extra Thick, 8.5cm x 9cm | 50 Sheets |
59.50€ Pack of 50 |
|
11766446 |
Thermo Scientific™
88615 |
Pierce Western Blotting Filter Paper, Extra Thick, 8cm x 13.5cm | 50 Sheets |
62.00€ Pack of 50 |
|
10675935 |
Thermo Scientific™
88620 |
Pierce Western Blotting Filter Paper, Extra Thick, 20cm x 20cm | 50 Sheets |
74.00€ Pack of 50 |
|
15105214 |
Thermo Scientific™
84783 |
Pierce Western Blotting Filter Paper, 7cm x 8.4cm | 100 Sheets |
65.00€ 100 sheets |
|
15115214 |
Thermo Scientific™
84784 |
Pierce Western Blotting Filter Paper, 8cm x 13.5cm | 100 Sheets |
70.75€ 100 sheets |
from Supplier 20-03-2020 |
Description
Sheets of blotting paper are necessary components of transfer sandwiches and cassettes that typically must be assembled for various kinds of protein or nucleic acid, gel-to-membrane, transfer protocols. High-quality Pierce Western Blotting Filter Papers are available as standard-thickness and extra-thick sheets that are pre-cut for convenience with mini gels.Highlights:
High-quality – clean cotton cellulose fiber paper manufactured with additive-free ultrapure water to eliminate sources of background signal and artifacts
Easy to use – pre-cut filter paper sheets in several convenient sizes for use with most mini gel sizes, tank transfer cassettes and semi-dry blotters
Two thicknesses – choose standard-thickness paper for traditional procedures or extra-thick filter paper for high capacity blotting or to eliminate using multiple sheets
Validated – tested and recommmend for use with protein methods, including the Thermo Scientific Pierce Fast Semi-Dry Blotter (Part No. 88217)