Learn More
Thermo Scientific™ Pierce™ Maleic Anhydride (Amine-binding) Plates
Bind amine-containing peptides and other molecules to microplate wells for use in binding assays with these maleic anhydride-activated plates.
171.00€ - 780.00€
Specifications
Binding Target Amine
Sufficient For Binding approx. 125pmol biotin-pentylamine per well
View More Specs
Products 4
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
10503544 View Documents Thermo Scientific™
15100
Pierce Maleic Anhydride Activated Plates, Clear, 96-Well Strip 5 plates 171.00€
Pack of 5 View alternative products
Save Up to 
»
EU Stock 3
Add to basket
10379253 View Documents Thermo Scientific™
15108
Pierce Maleic Anhydride Activated Plates, White, 96-Well 5 plates 175.00€
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-09-2019
Add to basket
10115383 View Documents Thermo Scientific™
15110
Pierce Maleic Anhydride Activated Plates, Clear, 96-Well 5 plates 183.00€
Pack of 5 View alternative products
Save Up to 
»
EU Stock 10
Add to basket
10712794 View Documents Thermo Scientific™
15112
Pierce Maleic Anhydride Activated Plates, Clear, 96-Well 5 x 5 plates 780.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 2
Add to basket
Description
Description
Pierce Maleic Anhydride Activated Plates are useful for immobilizing peptides and other ligands that do not coat efficiently by passive adsorption. Reaction of the maleic anhydride groups with primary amines (–NH2) results in formation of amide bonds that are stable at neutral pH and above. Acidic conditions hydrolyze the bond; for example, the half-life of hydrolysis at pH 3.5 and 37°C is approximately 11 hours. Therefore, coating reactions are best performed at pH 8 to 9, and the reacted plates are best used at pH greater than 7 for ELISA and other methods.
Highlights:
  • Spontaneously react with primary amines
  • A direct, simple approach to covalently attach molecules to the well surfaces of assay plates
  • Maleic anhydride maintains its integrity and availability for coupling for months under dry, room temperature conditions
Specifications
Specifications
Amine
Binding approx. 125pmol biotin-pentylamine per well