missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ In-Line Caps for 1.1 mL Microtube System
Pre-racked 1.1mL-tall microtubes boxed to match a standard 96-well microplate footprint for multichannel pipetting or automated workstations.
45.68€ - 120.00€
| Product Code | Brand | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Price | Quantity & Availability | |||||
| 10753374 |
Thermo Scientific™
15082 |
Pierce 1.1mL Microtube Racked System |
120.00€ Pack of 10 |
|
10593534 |
Thermo Scientific™
15086 |
Pierce 1.1mL Microtubes |
69.75€ Pack of 960 |
|
10464825 |
Thermo Scientific™
15088 |
Pierce 1.1mL Microtube In-Line Caps |
45.68€ Pack of 120 |
|
Description
The microtubes resembles 1mL pipette tips except that they have round bottoms. Like a box of 1mL pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.Highlights:
- 8 x 12 format simplifies and speeds applications in which a multi-channel pipettor is used
- Format is also compatible with robotic workstations
- Thermo Scientific Microtube System can be centrifuged