Learn More
WTW™ photoLab™ S12 Filter Photometers
Ideal for comprehensive routine measuring operations in wastewater and drinking water, food industry as well as in service and teaching labs. The photoLab™ S12 filter photometer with 12 wavelengths combines lab precision with highest comfort and highest speeds for extensive water analytics
Brand: WTW™ 250024D
Additional Details : Weight : 2.70900kg
Description
- The photoLab™ S12 filter photometer with 12 wavelengths combines lab precision with highest comfort and highest speeds for extensive water analytics
- photoLab™ S12 with reference beam optics has 12 wavelengths for quick tests in 16 mm round and rectangular cuvettes.
- Barcode detection for round and rectangular cuvettes
- AQA/IQC multi-level
- User-defined methods, easy to operate and maintenance-free
- Maximum comfort at minimal costs! Just plug in the cuvette, read measured value, done
Specifications
LCD | |
10.62 in. | |
5.51 in. | |
50 Hz | |
340, 410, 445, 500, 525, 550, 565, 605, 620, 665, 690, 820nm | |
10.23 in. |
±2nm | |
270 mm | |
230 V | |
140 mm | |
260 mm |