missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phorbol 12-Myristate 13-Acetate (Clear or Whitish Film), Fisher BioReagents™
Click to view available options
Quantity:
1 mg
Packaging:
Glass Ampule
Description
Phorbol 12-Myristate 13-Acetate (Clear or Whitish Film)
PMA is an activator of Protein Kinase C; it is a skin irritant and mouse skin tumor promoter.
Specifications
Melting Point | 72°C |
Color | White |
Assay Percent Range | ≥99.0 % |
Packaging | Glass Ampule |
Quantity | 1 mg |
Formula Weight | 616.8 |
Percent Purity | 99.0% min. |
Physical Form | Solid |
Chemical Name or Material | Phorbol 12-Myristate 13-Acetate |
Safety and Handling
Product Identifier
- Phorbol 12-myristate 13-acetate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'