missing translation for 'onlineSavingsMsg'
Learn More

Phenoxazine, 97%, Thermo Scientific Chemicals

Catalog Number p-3736605
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
135-67-1
C12H9NO
183.21
MFCD00005014
TZMSYXZUNZXBOL-UHFFFAOYSA-N
phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
67278
10H-phenoxazine
N1C2=CC=CC=C2OC2=CC=CC=C12
This item is not returnable. View return policy
135-67-1
C12H9NO
183.21
MFCD00005014
TZMSYXZUNZXBOL-UHFFFAOYSA-N
phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
67278
10H-phenoxazine
N1C2=CC=CC=C2OC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 154.0°C to 159.0°C
Color Gray to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 27, 62
Merck Index 15, 7365
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in abs.methanol, ethanol, ether,, chloroform and benzene, sparingly soluble in petroleum ether
Formula Weight 183.21
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenoxazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'