missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, U.S.P./N.F. grade, MP Biomedicals™

Product Code. 11428240 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g

Product Code. 11428240

Brand: MP Biomedicals 0210260883

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym 3, 3-bis[4-Hydroxyphenyl]-1-[3H]- isobenzofuranone
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Assay Percent Range U.S.P./N.F. grade
Formula Weight 318.3
Density 1.29g/cm3
Color White to White/Yellow
Quantity 250 g
Physical Form Crystals
Chemical Name or Material Phenolphthalein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.