Learn More
MP Biomedicals™ Phenol:Chloroform Saturated Solution
This is a ready-to-use saturated solution of phenol and chloroform suitable for the extraction and purification of DNA. This solution is supplied at pH 6.7, and a bottle of alkaline buffer is included to adjust the pH to 8.0 if desired. 1:1 solution
Brand:  MP Biomedicals™ 04802521
Additional Details : CAS : 108-95-2 Weight : 0.01000kg Transport : UN number : 2821 Chem class : 6.1 Pack group : II
Product Code. 11411602
  • 326.00€ / 400mL
Estimated Shipment
21-04-2020
Add to basket
Description
Description
  • More convenient and safer than preparing phenol solutions from crystals.
  • Premixed
  • Ready-to-use solution
Specifications
Specifications
Phenol:Chloroform Saturated Solution
400 mL