missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ PharMed™ BPT L/S™ 2-Stop Pump Tubing
Protect your light-sensitive samples
Brand: Masterflex™ 96114-14
Additional Details : Weight : 0.80000kg
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
- Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable
- For use with Masterflex™ multichannel pump heads.
Specifications
Beige | |
40 psi | |
1.6 mm | |
1.25 ft. | |
Stopped Pump Tubing | |
-20°C | |
14 | |
1/16 in. |
100°C | |
Yes | |
PharMed BPT | |
0.38 m | |
Cell and Tissue Culture Work, Pharmaceutical, Light-Sensitive Samples | |
8 | |
130 mL/min |