missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, ACS Reagent, 99%, Honeywell Fluka™

Catalog Number p-7120655
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
10450-60-9
H5IO6
227.94
MFCD00011344
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
O[I](O)(O)(O)(O)=O
This item is not returnable. View return policy
10450-60-9
H5IO6
227.94
MFCD00011344
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
O[I](O)(O)(O)(O)=O

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Appearance Powder or Crystals
Color White
Melting Point 122°C
Packaging Glass Bottle
Quantity 25 g
UN Number UN3085
Formula Weight 227.94g/mol
Percent Purity 99%
Chemical Name or Material Periodic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'