missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Periodic Acid, ACS Reagent, 99%, Honeywell™ Fluka™

ACS Reagent, 99%

Brand:  Honeywell Fluka™ 375810-100G

Additional Details : CAS : 10450-60-9 Weight : 0.10000kg Transport : UN number : 3085 Chem class : 5.1 Pack group : II

Packaging Glass Bottle
Quantity 100g

 View more versions of this product

Product Code. 15625150

  • 232.58€ / 100g


EU Stock 7
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10450-60-9
227.938
TWLXDPFBEPBAQB-UHFFFAOYSA-N
25289
pentahydroxy(oxo)-$l^{7}-iodane
H5IO6
MFCD00011344
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
CHEBI:29150
OI(=O)(O)(O)(O)O

Specifications

Periodic acid
White
10450-60-9
H5IO6
UN3085
TWLXDPFBEPBAQB-UHFFFAOYSA-N
pentahydroxy(oxo)-$l^{7}-iodane
25289
227.94g/mol
≤0.01%
Glass Bottle
122°C
As SO4, 0 to 100mg/kg
100g
Powder or Crystals
MFCD00011344
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
OI(=O)(O)(O)(O)O
227.938
CHEBI:29150
99%
0 to 30mg/kg
0 to 0.01%
Videos
SDS Documents