Learn More
Brand™ PCR Plate Combo Pack PROMO
Provides additional savings than when plates and fiilm are purchased individually. Brand™ PCR Plate Combo Pack PROMO includes 96-well PCR plates available in different skirt and profile types along with high quality qPCR sealing film.
263.00€
Specifications
Quantity | 50/Pk. |
---|---|
No. of Wells | 96 |
Format | 96-well |
Product Type | PCR Combo Pack |
Includes: 96-well PCR plates, PCR-sealing film
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15504419
|
Brand™
781231 |
Non-skirted PCR plates |
263.00€
1 set |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15514419
|
Brand™
781232 |
Elevated rim low profile plates |
263.00€
1 set |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15524419
|
Brand™
781233 |
Semi-Skirted, Elevated, Low profile plates |
263.00€
1 set |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
15534419
|
Brand™
781234 |
Standard profile plates |
263.00€
1 set |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15594409
|
Brand™
781230 |
Non-Skirted, Low profile plates |
263.00€
1 set |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Premium PCR/qPCR transparent plates- 96-well
- Skirted, non-skirted, or semi-skirted
- Low, standard or elevated rim profile
- Ultra-thin walled for quick and constant thermal transfer
- Free of DNase, DNA, RNase and endotoxins
- No warping, no wiggling, suitable for automation
- Highly transparent for qPCR
- Press to seal
- Convenient end tabs
- No annoying adhesive residues
Specifications
50/Pk. | |
96 | |
96-well | |
PCR Combo Pack |