missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Parthenolide
Click to view available options
Quantity:
100 mg
500 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 115.0°C |
Color | Clear |
Quantity | 100 mg |
Merck Index | 15, 7152 |
Solubility Information | Solubility aq. soln.: .. Other solubilities: soluble in chloroform |
Formula Weight | 248.32 |
Chemical Name or Material | Parthenolide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'