Learn More
Uvex™ Outdoor Jacket 7408-Moonless Night
Catalog Number 17210392
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
The uvex suXXeed collection gets everything spot on: well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove. Can be worn in industrial and metal processing settings as well as for leisure
  • Material: 100% polyamid with PU-finish inside jacket: 100% polyester
  • Micro fleece 3 in 1 functional jacket in breathable, wind and waterproof rep-top-material.
  • Ideal for applications in the winter and for the in-between seasons.
  • Contrasting colored inserts on shoulder, sleeves and bodice, reflective piping on front and back side for better visibility, zip-in fleece jacket with 3-points- fixation, combination of gliding lining and ventilating mesh lining, much storage spaces trough side pockets
  • Napoleonpocket and inside pocket with zip fastener and inside mobile phone pocket, hood is able to stow in collar, comfortable collar lining in fleece, in front concealed 2-way zip fastener with chin protection facing, elastic seamed sleeves with Velcro fastener, width-adjustable, seam with elastic draw-string inside jacket: fleece jacket separately to wear
  • 2 side pocket and inside chest pocket left, 1 chest pocket with zip fastener
  • Sizes: S XXXL
  • Care instructions: 30°C washable (gentle wash), impossible are chlorinate, ironing, dry cleaning and tumble dryer
Size 4X-Large
Gender Men's
Color Gray
Closure Type Zipper, Press Studs
Disposable No
Thickness 145 g/m²