missing translation for 'onlineSavingsMsg'
Learn More

o-Cresolphthalein Complex, Honeywell Fluka™

Catalog Number 15626100
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
2411-89-4
C32H32N2O12
636.61
MFCD00005911
IYZPEGVSBUNMBE-UHFFFAOYSA-N
75485
2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxymethyl)amino]acetic acid
CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
This item is not returnable. View return policy
2411-89-4
C32H32N2O12
636.61
MFCD00005911
IYZPEGVSBUNMBE-UHFFFAOYSA-N
75485
2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxymethyl)amino]acetic acid
CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 179°C to 182°C
Boiling Point >260°C
UN Number NONH for all modes of transport
Beilstein 381994
Formula Weight 636.6g/mol
Odor Unpleasant
Packaging Glass Bottle
Color Yellow
Quantity 1 g
Physical Form Powder
Chemical Name or Material o-Cresolphthalein Complexone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'